Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCARF2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547339
Description
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human SREC-I is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
The protein encoded by this gene is similar to SCARF1/SREC-I, a scavenger receptor protein that mediates the binding and degradation of acetylated low density lipoprotein (Ac-LDL). This protein has only little activity of internalizing modified low density lipoproteins (LDL), but it can interact with SCARF1 through its extracellular domain. The association of this protein with SCARF1 is suppressed by the presence of scavenger ligands. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
SCARF2 | |
Polyclonal | |
Unconjugated | |
SCARF2 | |
AI194363; NSR1; Scarf1; SCARF2; Scatf1; Scavenger receptor class F member 2; scavenger receptor class F, member 1; scavenger receptor class F, member 2; scavenger receptor expressed by endothelial cells 2 (acetyl LDL receptor like); scavenger receptor expressed by endothelial cells 2 protein; scavenger receptor type F; SREC2; SrecII; SREC-II; SRECRP-1; SREPCR; VDEGS; W53987; W65560 | |
Goat | |
Antigen Affinity Chromatography | |
RUO | |
91179 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q96GP6 | |
SCARF2 | |
Mouse myeloma cell line NS0-derived recombinant human SREC-II/SCARF2 Ala42-Gly442. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction