Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCARF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557146
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557146 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557146 Supplier Invitrogen™ Supplier No. PA557146
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HHDLDNTLNC SFLEPPSGLE QPSPSWSSRA SFSSFDTTDE GPVYCVPHEE APAESRDPEV PTVP Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 89%.

The protein encoded by this gene is similar to SCARF1/SREC-I, a scavenger receptor protein that mediates the binding and degradation of acetylated low density lipoprotein (Ac-LDL). This protein has only little activity of internalizing modified low density lipoproteins (LDL), but it can interact with SCARF1 through its extracellular domain. The association of this protein with SCARF1 is suppressed by the presence of scavenger ligands. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCARF2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCARF2
Gene Accession No. P59222, Q96GP6
Gene Alias AI194363; NSR1; Scarf1; SCARF2; Scatf1; Scavenger receptor class F member 2; scavenger receptor class F, member 1; scavenger receptor class F, member 2; scavenger receptor expressed by endothelial cells 2 (acetyl LDL receptor like); scavenger receptor expressed by endothelial cells 2 protein; scavenger receptor type F; SREC2; SrecII; SREC-II; SRECRP-1; SREPCR; VDEGS; W53987; W65560
Gene Symbols SCARF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCARF2. Recombinant protein control fragment (Product #RP-95366).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224024, 287949, 91179
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.