Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scavenger Receptor BI/BII Polyclonal Antibody, Biotin, Bioss
SDP

Catalog No. 502279929
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-9929 Supplier Bioss Supplier No. BS17277RBIOTIN
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Scavenger Receptor BI/BII Polyclonal antibody specifically detects Scavenger Receptor BI/BII in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen Scavenger Receptor BI/BII
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene SCARB2
Gene Accession No. O35114, Q14108, Q61009, Q8WTV0
Gene Alias 85 kDa lysosomal membrane sialoglycoprotein; 85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2; 9330185J12Rik; AI120173; AMRF; CD36; CD36 and LIMPII analogous 1; CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 (scavanger receptor class B type 1); CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2; CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2 (lysosomal integral membrane protein II); CD36 antigen-like 1; CD36 antigen-like 2; CD36L1; CD36L2; Cla1; CLA-1; Collagen type I receptor, thrombospondin receptor-like 1; D5Ertd460e; EPM4; HDL QTL 1; Hdlq1; HDLQTL6; Hlb398; HLGP85; LGP85; LIMP II; LIMP2; LIMP-2; LIMPII; lysosome membrane protein 2; Lysosome membrane protein II; MLGP85; mSR-BI; Scarb1; Scarb2; scavanger receptor class B type 1; scavenger receptor b1; scavenger receptor class B member 1; Scavenger receptor class B member 2; scavenger receptor class B type I; scavenger receptor class B type III; scavenger receptor class B, member 1; scavenger receptor class B, member 2; scavenger receptor class B1; SRB1; SR-B1; SR-B1/B2; SRBI; SR-BI; SR-BI/BII; SR-BII
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Scavenger Receptor BI + BII (31-130).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12492, 20778, 949, 950
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.