Learn More
SCC-112 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300088A
Description
The recommended shelf life for this product is 1 year from date of receipt.
PDS5A belongs to the PDS5 family. It contains 1 HEAT repeat and 3 LRR (leucine-rich) repeats. PDS5A may contribute to regulation of sister chromatid cohesion during mitosis.Specifications
| SCC-112 | |
| Polyclonal | |
| Unconjugated | |
| PDS5A | |
| cell proliferation-inducing gene 54 protein, KIAA0648, PDS5, regulator of cohesion maintece, homolog A, PDS5, regulator of cohesion maintece, homolog A (S. cerevisiae), PIG54, SCC112, SCC-112, sister chromatid cohesion protein 112 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23244 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q29RF7 | |
| PDS5A | |
| Between 1150 and 1200 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.