Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCC112 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100425C
Description
SCC112 Polyclonal antibody specifically detects SCC112 in Human samples. It is validated for Western BlotSpecifications
SCC112 | |
Polyclonal | |
Western Blot | |
Cell proliferation-inducing gene 54 protein, DKFZp686B19246, FLJ41012, KIAA0648MGC131948, PDS5, PDS5, regulator of cohesion maintenance, homolog A (S. cerevisiae), PIG54, SCC-112MGC161503, Sister chromatid cohesion protein 112, sister chromatid cohesion protein PDS5 homolog A | |
A synthetic peptide mapping to a region between residues 1150 and 1200 of human SCC-112 using the numbering given in entry NP_056015.1 (GeneID 23244). | |
0.1 mL | |
Cell Cycle and Replication, DNA Repair | |
23244 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction