Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCC112 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100426
Description
SCC112 Polyclonal specifically detects SCC112 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
SCC112 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
PDS5A | |
A synthetic peptide mapping to a region between residues 1250 and 1297 (C-terminus) of human SCC-112 using the numbering given in entry NP_056015.1 (GeneID 23244). | |
100 μL | |
Cell Cycle and Replication, DNA Repair | |
23244 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:10000-1:25000, Immunoprecipitation 1-4 ug/mg of lysate | |
Cell proliferation-inducing gene 54 protein, DKFZp686B19246, FLJ41012, KIAA0648MGC131948, PDS5, PDS5, regulator of cohesion maintenance, homolog A (S. cerevisiae), PIG54, SCC-112MGC161503, Sister chromatid cohesion protein 112, sister chromatid cohesion protein PDS5 homolog A | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction