Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCCPDH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102589
Description
Antibody detects endogenous levels of total SCCPDH.
SCCPDH (Probable saccharopine dehydrogenase) is a 429 amino acid protein that belongs to the saccharopine dehydrogenase family. The SCCPDH gene is conserved in chimpanzee, dog, cow, mouse, rat, chicken, fruit fly, mosquito and C. elegans, and maps to human chromosome 1q44. Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Stickler syndrome, Parkinson's, Gaucher disease and Usher syndrome are also associated with chromosome 1. A breakpoint has been identified in 1q which disrupts the DISC1 gene and is linked to schizophrenia. Aberrations in chromosome 1 are found in a variety of cancers including head and neck cancer, malignant melanoma and multiple myeloma.
Specifications
SCCPDH | |
Polyclonal | |
Unconjugated | |
SCCPDH | |
AW214504; C330023F11Rik; CGI 49; CGI49; CGI-49; hypothetical protein LOC507289; NET11; probable saccharopine dehydrogenase; RGD1311440; RP11-439E19.2; saccharopine dehydrogenase (putative); saccharopine dehydrogenase-like oxidoreductase; Sccpdh; SCPDH | |
Rabbit | |
Affinity chromatography | |
RUO | |
109232, 305021, 51097 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6AY30, Q8NBX0, Q8R127 | |
SCCPDH | |
A synthesized peptide derived from human SCCPDH(Accession Q8NBX0), corresponding to amino acid residues L130-F180. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction