Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCCPDH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531943
Description
Recommended positive controls: U87-MG. Predicted reactivity: Human (99%), Mouse (85%), Rat (87%), Pig (84%), Chimpanzee (98%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SCCPDH (Probable saccharopine dehydrogenase) is a 429 amino acid protein that belongs to the saccharopine dehydrogenase family. The SCCPDH gene is conserved in chimpanzee, dog, cow, mouse, rat, chicken, fruit fly, mosquito and C. elegans, and maps to human chromosome 1q44. Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Stickler syndrome, Parkinson's, Gaucher disease and Usher syndrome are also associated with chromosome 1. A breakpoint has been identified in 1q which disrupts the DISC1 gene and is linked to schizophrenia. Aberrations in chromosome 1 are found in a variety of cancers including head and neck cancer, malignant melanoma and multiple myeloma.
Specifications
SCCPDH | |
Polyclonal | |
Unconjugated | |
SCCPDH | |
AW214504; C330023F11Rik; CGI 49; CGI49; CGI-49; hypothetical protein LOC507289; NET11; probable saccharopine dehydrogenase; RGD1311440; RP11-439E19.2; saccharopine dehydrogenase (putative); saccharopine dehydrogenase-like oxidoreductase; Sccpdh; SCPDH | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51097 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8NBX0 | |
SCCPDH | |
Recombinant protein fragment corresponding to a region within amino acids 37 and 230 of Human SCCPDH. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction