Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCCPDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596070
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596070 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596070 Supplier Invitrogen™ Supplier No. PA596070
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATEQRPFHL VVFGASGFTG QFVTEEVARE QVDPERSSRL PWAVAGRSRE KLQRVLEKAA LKLGRPTLSS EVGIIICDIA NPASLDEMAK QATVVLNCVG PYRFYGEPVI KACIENGASC IDISGEPQFL ELMQLKYHEK AADKGVYIIG SSGFDSIPAD LGVIYTRNKM NGTLTAVESF LTIHSGPEGL SIHDGTWKSA IYGFGDQSNL RKLRNVSNLK PVPLIGPKLK RRWPISYCRE LKGYSIPFMG SDVSVVRRTQ RYLYENLEES; Positive Samples: THP-1, PC-3, 22Rv1, U-251MG, A673, Mouse heart.

SCCPDH (Probable saccharopine dehydrogenase) is a 429 amino acid protein that belongs to the saccharopine dehydrogenase family. The SCCPDH gene is conserved in chimpanzee, dog, cow, mouse, rat, chicken, fruit fly, mosquito and C. elegans, and maps to human chromosome 1q44. Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Stickler syndrome, Parkinson's, Gaucher disease and Usher syndrome are also associated with chromosome 1. A breakpoint has been identified in 1q which disrupts the DISC1 gene and is linked to schizophrenia. Aberrations in chromosome 1 are found in a variety of cancers including head and neck cancer, malignant melanoma and multiple myeloma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCCPDH
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SCCPDH
Gene Accession No. Q8NBX0, Q8R127
Gene Alias AW214504; C330023F11Rik; CGI 49; CGI49; CGI-49; hypothetical protein LOC507289; NET11; probable saccharopine dehydrogenase; RGD1311440; RP11-439E19.2; saccharopine dehydrogenase (putative); saccharopine dehydrogenase-like oxidoreductase; Sccpdh; SCPDH
Gene Symbols SCCPDH
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human SCCPDH (NP_0570862).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109232, 51097
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.