Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCD-1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP126393AF532
Description
SCD-1 Polyclonal specifically detects SCD-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SCD-1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
FADS5, MSTP008, SCD1, SCDOS, Stearoyl-CoA desaturase | |
Rabbit | |
Affinity Purified | |
Cancer, Diabetes Research, Lipid and Metabolism, Signal Transduction, Stem Cell Markers | |
6319.0 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
SCD | |
Synthetic peptide made to an internal portion of human SCD (within residues 50-100). [Swiss-ProtNo. O00767] | |
0.1 mL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction