Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517331
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517331 Supplier Invitrogen™ Supplier No. PA517331
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

Stearoyl-CoA desaturase (SCD; EC 1.14.99.5) is an iron-containing enzyme that catalyzes a rate-limiting step in the synthesis of unsaturated fatty acids. The principal product of SCD is oleic acid, which is formed by desaturation of stearic acid. The ratio of stearic acid to oleic acid has been implicated in the regulation of cell growth and differentiation through effects on cell membrane fluidity and signal transduction. Four SCD isoforms, Scd1 through Scd4, have been identified in mouse. In contrast, only 2 SCD isoforms, SCD1 and SCD5., have been identified in human. SCD1 shares about 85% amino acid identity with all 4 mouse SCD isoforms, as well as with rat Scd1 and Scd2. In contrast, SCD5 shares limited homology with the rodent SCDs and appears to be unique to primates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCD
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene Scd
Gene Accession No. O00767, P13516
Gene Alias AA589638; ab; acyl-CoA desaturase; Acyl-CoA desaturase 1; acyl-CoA desaturase 2; AI265570; asebia; delta(9)-desaturase; Delta(9)-desaturase 1; delta(9)-desaturase 2; Delta-9 desaturase; delta-9 desaturase 1; Delta-9 desaturase 2; delta-9-desaturase; FADS5; fatty acid desaturase; Fatty acid desaturase 1; fatty acid desaturase 2; HGNC:10571; hSCD1; MSTP008; OTTHUMP00000020280; predicted protein of HQ0998; PRO0998; PRO1933; Scd; Scd1; Scd-1; Scd2; SCDOS; stearoyl-CoA desaturase; stearoyl-CoA desaturase (delta-9-desaturase); stearoyl-CoA desaturase 1; Stearoyl-CoA desaturase 2; stearoyl-CoA desaturase opposite strand; stearoyl-Coenzyme A desaturase 1; stearoyl-Coenzyme A desaturase 2; stearyl-CoA desaturase
Gene Symbols Scd, Scd1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding M38 of human SCD1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20249, 6319
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.