Learn More
Invitrogen™ SCD Polyclonal Antibody

Description
It is not recommended to aliquot this antibody.
Specifications
Specifications
| Antigen | SCD |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 16 μg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 0.15M NaCl, 100 μg/mL BSA, 50% glycerol and no preservative; pH 7.5 |
| Gene | Scd |
| Gene Accession No. | O00767, P13516 |
| Gene Alias | AA589638; ab; acyl-CoA desaturase; Acyl-CoA desaturase 1; acyl-CoA desaturase 2; AI265570; asebia; delta(9)-desaturase; Delta(9)-desaturase 1; delta(9)-desaturase 2; Delta-9 desaturase; delta-9 desaturase 1; Delta-9 desaturase 2; delta-9-desaturase; FADS5; fatty acid desaturase; Fatty acid desaturase 1; fatty acid desaturase 2; HGNC:10571; hSCD1; MSTP008; OTTHUMP00000020280; predicted protein of HQ0998; PRO0998; PRO1933; Scd; Scd1; Scd-1; Scd2; SCDOS; stearoyl-CoA desaturase; stearoyl-CoA desaturase (delta-9-desaturase); stearoyl-CoA desaturase 1; Stearoyl-CoA desaturase 2; stearoyl-CoA desaturase opposite strand; stearoyl-Coenzyme A desaturase 1; stearoyl-Coenzyme A desaturase 2; stearyl-CoA desaturase |
| Gene Symbols | Scd, Scd1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.