Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559963
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559963 Supplier Invitrogen™ Supplier No. PA559963
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NTQHIQKEGR ALNQEAACEM LREWHQGHIL KVTLPGLHIL ALLHTHCNHS EKCCLMLRAL SVSLEVF Highest antigen sequence identity to the following orthologs: Mouse - 24%, Rat - 23%.

Stearoyl-CoA desaturase is an integral membrane protein of the endoplasmic reticulum that catalyzes the formation of monounsaturated fatty acids from saturated fatty acids. SCD may be a key regulator of energy metabolism with a role in obesity and dislipidemia. Four SCD isoforms, Scd1 through Scd4, have been identified in mouse. In contrast, only 2 SCD isoforms, SCD1 and SCD5, have been identified in human. SCD1 shares about 85% amino acid identity with all 4 mouse SCD isoforms, as well as with rat Scd1 and Scd2. In contrast, SCD5 shares limited homology with the rodent SCDs and appears to be unique to primates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCD5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCD5
Gene Accession No. Q86SK9
Gene Alias ACOD4; acyl-CoA-desaturase 4; FADS4; HSCD5; SCD2; SCD4; SCD5; Stearoyl-CoA 9-desaturase; stearoyl-CoA desaturase 2; stearoyl-CoA desaturase 4; stearoyl-CoA desaturase 5
Gene Symbols SCD5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCD5. Recombinant protein control fragment (Product #RP-90985).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79966
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.