Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546702
Description
Peptide sequence: HRAHHKYSET DADPHNARRG FFFSHIGWLF VRKHRDVIEK GRKLDVTDLL.
Stearoyl-CoA desaturase is an integral membrane protein of the endoplasmic reticulum that catalyzes the formation of monounsaturated fatty acids from saturated fatty acids. SCD may be a key regulator of energy metabolism with a role in obesity and dislipidemia. Four SCD isoforms, Scd1 through Scd4, have been identified in mouse. In contrast, only 2 SCD isoforms, SCD1 and SCD5, have been identified in human. SCD1 shares about 85% amino acid identity with all 4 mouse SCD isoforms, as well as with rat Scd1 and Scd2. In contrast, SCD5 shares limited homology with the rodent SCDs and appears to be unique to primates.
Specifications
| SCD5 | |
| Polyclonal | |
| Unconjugated | |
| SCD5 | |
| ACOD4; acyl-CoA-desaturase 4; FADS4; HSCD5; SCD2; SCD4; SCD5; Stearoyl-CoA 9-desaturase; stearoyl-CoA desaturase 2; stearoyl-CoA desaturase 4; stearoyl-CoA desaturase 5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79966 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86SK9 | |
| SCD5 | |
| Synthetic peptide directed towards the middle region of human SCD5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction