Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCEL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558908 Supplier Invitrogen™ Supplier No. PA558908
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KMSPTGNEMK STTQGTTRKQ QDFHEVNKRR TFLQDNSWIK KRPEEEKDEN YGRVVLNRHN SHDALDRKVN ERDVPKATIS RYS Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 78%.

May function in the assembly or regulation of proteins in the cornified envelope. The LIM domain may be involved in homotypic or heterotypic associations and may function to localize sciellin to the cornified envelope.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCEL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCEL
Gene Accession No. O95171
Gene Alias 9230114I02Rik; Scel; Sciellin
Gene Symbols SCEL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCEL. Recombinant protein control fragment (Product #RP-97961).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8796
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.