Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ SCF (C-Kit Ligand) Recombinant Human Protein

Catalog No. p-7076224
Encompass_Preferred
Click to view available options
Quantity:
1 mg

SCF (C-Kit Ligand); Recombinant Human Protein

Cell Therapy Systems CTS™ Recombinant Human Stem Cell Factor (SCF) is a bioactive protein intended for use in cell culture applications, including pre-clinical and clinical cell therapy studies. SCF is a pleiotropic factor that acts in utero in germ cell and neural cell development and hematopoiesis. These activities are all believed to reflect a role in cell migration. In adults, it functions pleiotropically, while mostly noted for its continued requirement in hematopoiesis. The lyophilized, E. coli-expressed SCF CTS™ Recombinant Human is verified for activity through dose-dependent proliferation of human MO-7e cells.

  • Qualified—helps reduce your burden in qualifying reagents for regulatory submission in cell therapy applications
  • Highly Pure—minimizes interference from other proteins or contaminants
  • Active—get more results with less protein

Simplified Transition into Clinical Research

GIBCO™ CTS™ products are of high quality and supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate. The GIBCO™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.

High Purity

To help verify that GIBCO™ CTS™ Recombinant growth factors are of the highest quality, each protein is analyzed for purity, along with structural homogeneity, to help ensure a biologically active protein. The purity of each GIBCO™ CTS™ recombinant growth factor is verified by SDS-PAGE to be over 95% and to have endotoxin levels below 0.1 ng/μg.

High Biological Activity

All GIBCO™ CTS™ Recombinant growth factors are verified for activity. Each lot has its own ED50 determined using an activity assay based on a specific function of the growth factor. The activity of our SCF CTS™ Recombinant Human growth factor is determined by the dose-dependent proliferation of human MO-7e cells to have an ED50 range of 2 to 10 ng/mL (specific activity: 5 × 105–1 × 105 units/mg). Optimal concentration for individual applications should be determined by a dose response assay.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Molecular Weight (g/mol) 18.5 kDa
Purification Method Sequential Chromatography
Quantity 1 mg
Source Human
Gene Alias SCF (c-kit Ligand)
Protein Length 164 aa
Species Human
Recombinant Recombinant
Content And Storage 2°C to 8°C
Classification Carrier-Free
Activity 2 ng/ml
Bioassay Proliferation of MO7e cells
Endotoxin Level <0.1 ng/μg
Shipping Condition Wet Ice
Expression System E. coli
Protein Family Growth Factors & Receptors
Protein Form Full Length, Recombinant, Ligand
Form Lyophilized
Protein Subtype Other Proteins
Research Category Clinical Research, Stem Cell Research, Differentiation, Oncology
Product Line Gibco™
Purity or Quality Grade 95 %
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.