Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCFD1 Antibody (OTI5G4), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274004JF549
Description
SCFD1 Monoclonal specifically detects SCFD1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SCFD1 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SCFD1 | |
Full length recombinant protein of human SCFD1 (NP_057190) produced in HEK293T cells. | |
0.1 mL | |
23256.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI5G4 | |
Western Blot, Immunohistochemistry | |
C14orf163, KIAA0917chromosome 14 open reading frame 163, sec1 family domain containing 1, sec1 family domain-containing protein 1, SLY1, SLY1 homolog, sly1p, STXBP1L2RA410, Syntaxin-binding protein 1-like 2, vesicle transport-related protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction