Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCFD1 Monoclonal Antibody (OTI1F4), TrueMAB™, OriGene
SDP

Catalog No. 502540170
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0170 30 μL
50-167-5324 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0170 Supplier OriGene Supplier No. TA504691S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SCFD1 Monoclonal antibody specifically detects SCFD1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SCFD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TBCC
Gene Accession No. Q15814
Gene Alias 3110021P21Rik; C14orf163; FKSG23; KIAA0917; Ra410; rSly1; Scfd1; Sec1 family domain containing 1; sec1 family domain-containing protein 1; Sly1; SLY1 homolog; Sly1p; STXBP1L2; Syntaxin-binding protein 1-like 2; vesicle transport-related protein; vesicle transport-related protein Ra410
Host Species Mouse
Immunogen Full length human recombinant protein of human TBCC produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6903
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.