Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCFD1 Monoclonal Antibody (OTI4D2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525962
Description
SCFD1 Monoclonal Antibody for Western Blot, IHC (P)
The de novo synthesis of pyrimidine nucleotides is required for mammalian cells to proliferate. This gene encodes a trifunctional protein which is associated with the enzymatic activities of the first 3 enzymes in the 6-step pathway of pyrimidine biosynthesis: carbamoylphosphate synthetase (CPS II), aspartate transcarbamoylase, and dihydroorotase. This protein is regulated by the mitogen-activated protein kinase (MAPK) cascade, which indicates a direct link between activation of the MAPK cascade and de novo biosynthesis of pyrimidine nucleotides.
Specifications
SCFD1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8WVM8 | |
SCFD1 | |
Full length human recombinant protein of SCFD1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4D2 | |
Unconjugated | |
SCFD1 | |
3110021P21Rik; C14orf163; FKSG23; KIAA0917; Ra410; rSly1; Scfd1; Sec1 family domain containing 1; sec1 family domain-containing protein 1; Sly1; SLY1 homolog; Sly1p; STXBP1L2; Syntaxin-binding protein 1-like 2; vesicle transport-related protein; vesicle transport-related protein Ra410 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23256 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction