Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCFD1 Mouse anti-Human, Clone: OTI10D8, lyophilized, TrueMAB™
SDP

Catalog No. 501668835
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-8835 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-8835 Supplier OriGene Supplier No. CF808410
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

SCFD1 is involved in vesicular transport between the endoplasmic reticulum and the Golgi.

Specifications

Antigen SCFD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI10D8
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene SCFD1
Gene Accession No. Q8WVM8
Gene Alias C14orf163, RA410, SLY1, SLY1P, STXBP1L2
Gene Symbols SCFD1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 66-190 of human SCFD1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23256
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.