Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCFD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598694
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598694 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598694 Supplier Invitrogen™ Supplier No. PA598694
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCFD1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The de novo synthesis of pyrimidine nucleotides is required for mammalian cells to proliferate. This gene encodes a trifunctional protein which is associated with the enzymatic activities of the first 3 enzymes in the 6-step pathway of pyrimidine biosynthesis: carbamoylphosphate synthetase (CPS II), aspartate transcarbamoylase, and dihydroorotase. This protein is regulated by the mitogen-activated protein kinase (MAPK) cascade, which indicates a direct link between activation of the MAPK cascade and de novo biosynthesis of pyrimidine nucleotides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCFD1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SCFD1
Gene Accession No. Q8WVM8
Gene Alias 3110021P21Rik; C14orf163; FKSG23; KIAA0917; Ra410; rSly1; Scfd1; Sec1 family domain containing 1; sec1 family domain-containing protein 1; Sly1; SLY1 homolog; Sly1p; STXBP1L2; Syntaxin-binding protein 1-like 2; vesicle transport-related protein; vesicle transport-related protein Ra410
Gene Symbols SCFD1
Host Species Rabbit
Immunogen Recombinant Human Sec1 family domain-containing protein 1 protein (373-642AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23256
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.