Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121140 Supplier Invitrogen™ Supplier No. PA5121140
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 22Rv1, HepG2, Mouse brain, Mouse lung, Mouse heart, Rat brain, Rat lung, Rat heart. The target is usually found in the following locations: Cytoplasmic vesicle, Peripheral membrane protein, Secreted, secretory vesicle lumen, secretory vesicle membrane. Immunogen sequence: KLISKEANNY EEDPNKPTSW TENQAGKIPE KVTPMAAIQD GLAKGENDET VSNTLTLTNG LERRTKTYSE DNFEELQYFP NFYALLKSID SEKEAKEKET LITIMKTLID FVKMMVKYGT ISPEEGVSYL ENLDEMIALQ TKNKLEKNAT DNISKLFPAP SEKSHEETDS TKEEAAKMEK EYGSLKDSTK DDNSNPGGKT DEPKGKTEAY LEAIRKNIEW LKKHDKKGNK EDYDLSKMRD FINKQADAYV EKGILDKEEA EAIKRIYSSL.

The protein encoded by this gene is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. Granins may serve as precursors for biologically active peptides. Some granins have been shown to function as helper proteins in sorting and proteolytic processing of prohormones; however, the function of this protein is unknown. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCG3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SCG3
Gene Accession No. P47867, P47868, Q8WXD2
Gene Alias 1B1075; AI385542; Chgd; Scg3; Secretogranin III; secretogranin-3; SgIII; UNQ2502/PRO5990
Gene Symbols SCG3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 199-468 of human SCG3 (NP_037375.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116635, 20255, 29106
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.