Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCG3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550259
Description
The antibody detects endogenous levels of total SCG3 protein.
The protein encoded by this gene is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. Granins may serve as precursors for biologically active peptides. Some granins have been shown to function as helper proteins in sorting and proteolytic processing of prohormones; however, the function of this protein is unknown. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
SCG3 | |
Polyclonal | |
Unconjugated | |
SCG3 | |
1B1075; AI385542; Chgd; Scg3; Secretogranin III; secretogranin-3; SgIII; UNQ2502/PRO5990 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20255, 29106 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P47867, Q8WXD2 | |
SCG3 | |
Fusion protein corresponding to a region derived from internal residues of human Secretogranin III. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction