Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579955
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579955 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579955 Supplier Invitrogen™ Supplier No. PA579955
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HELA Cell, Marker 1113, rat brain tissue, rat kidney tissue, rat PC-12 whole cellmouse brain tissue, mouse kidney tissue, mouse pancreas tissue, mouse NEURO-2A whole cell. IHC: Rat Brain Tissue, Human Rectal Cancer Tissue.

The protein encoded by this gene is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. Granins may serve as precursors for biologically active peptides. Some granins have been shown to function as helper proteins in sorting and proteolytic processing of prohormones; however, the function of this protein is unknown. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCG3
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene SCG3
Gene Accession No. P47867, P47868, Q8WXD2
Gene Alias 1B1075; AI385542; Chgd; Scg3; Secretogranin III; secretogranin-3; SgIII; UNQ2502/PRO5990
Gene Symbols SCG3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human Secretogranin 3 (438-450aa DFINKQADAYVEK).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116635, 20255, 29106
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.