Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCG5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570631
Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 85%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 86%; Rat: 93%.
Acts as a molecular chaperone for PCSK2/PC2, preventing its premature activation in the regulated secretory pathway. Binds to inactive PCSK2 in the endoplasmic reticulum and facilitates its transport from there to later compartments of the secretory pathway where it is proteolytically matured and activated. Also required for cleavage of PCSK2 but does not appear to be involved in its folding. Plays a role in regulating pituitary hormone secretion. The C-terminal peptide inhibits PCSK2 in vitro.
Specifications
SCG5 | |
Polyclonal | |
Unconjugated | |
Scg5 | |
7B2; AI325031; C-terminal peptide; hypothetical protein MGC73132; LOC508224 protein; Neuroendocrine protein 7B2; neuronal endocrine protein; N-terminal peptide; P SGNE1; P7B2; pituitary polypeptide; prohormone convertase chaperone; SCG5; Secretogranin V; secretogranin V (7B2 protein); secretogranin-5; Secretory granule endocrine protein I; secretory granule neuroendocrine protein 1; secretory granule neuroendocrine protein 1, 7B2 protein; Secretory granule neuroendocrine, protein 1 (7B2 protein); secretory granule, neuroendocrine protein 1; secretory granule, neuroendocrine protein 1 (7B2 protein); SGNE1; Sgne-1; SgV; zgc:73132 | |
Rabbit | |
Affinity chromatography | |
RUO | |
6447 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P05408 | |
Scg5 | |
synthetic peptide directed towards the middle region of human SCG5 (aa 148-197). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction