Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCG5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103965 Supplier Invitrogen™ Supplier No. PA5103965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SCG5.

Acts as a molecular chaperone for PCSK2/PC2, preventing its premature activation in the regulated secretory pathway. Binds to inactive PCSK2 in the endoplasmic reticulum and facilitates its transport from there to later compartments of the secretory pathway where it is proteolytically matured and activated. Also required for cleavage of PCSK2 but does not appear to be involved in its folding. Plays a role in regulating pituitary hormone secretion. The C-terminal peptide inhibits PCSK2 in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCG5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Scg5
Gene Accession No. P05408, P12961, P27682
Gene Alias 7B2; AI325031; C-terminal peptide; hypothetical protein MGC73132; LOC508224 protein; Neuroendocrine protein 7B2; neuronal endocrine protein; N-terminal peptide; P SGNE1; P7B2; pituitary polypeptide; prohormone convertase chaperone; SCG5; Secretogranin V; secretogranin V (7B2 protein); secretogranin-5; Secretory granule endocrine protein I; secretory granule neuroendocrine protein 1; secretory granule neuroendocrine protein 1, 7B2 protein; Secretory granule neuroendocrine, protein 1 (7B2 protein); secretory granule, neuroendocrine protein 1; secretory granule, neuroendocrine protein 1 (7B2 protein); SGNE1; Sgne-1; SgV; zgc:73132
Gene Symbols Scg5
Host Species Rabbit
Immunogen A synthesized peptide derived from human SCG5(Accession P05408), corresponding to amino acid residues Q111-G161.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20394, 25719, 6447
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.