Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCG5 Polyclonal Antibody

SCG5 Polyclonal Antibody
Supplier: Thermo Scientific PA568244
Description
SCG5 Polyclonal Antibody for Western Blot

Specifications
SCG5 | |
Polyclonal | |
Unconjugated | |
Scg5 | |
7B2; AI325031; C-terminal peptide; hypothetical protein MGC73132; LOC508224 protein; Neuroendocrine protein 7B2; neuronal endocrine protein; N-terminal peptide; P SGNE1; P7B2; pituitary polypeptide; prohormone convertase chaperone; SCG5; Secretogranin V; secretogranin V (7B2 protein); secretogranin-5; Secretory granule endocrine protein I; secretory granule neuroendocrine protein 1; secretory granule neuroendocrine protein 1, 7B2 protein; Secretory granule neuroendocrine, protein 1 (7B2 protein); secretory granule, neuroendocrine protein 1; secretory granule, neuroendocrine protein 1 (7B2 protein); SGNE1; Sgne-1; SgV; zgc:73132 | |
Rabbit | |
affinity chromatography | |
RUO | |
20394, 25719, 6447 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P05408, P12961, P27682 | |
Scg5 | |
Synthesized peptide derived from human SCG5. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction