Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCGB1D2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550697

Catalog No. PA550697


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total SCGB1D2 protein.

May bind androgens and other steroids, may also bind estramustine, a chemotherapeutic agent used for prostate cancer. May be under transcriptional regulation of steroid hormones.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SCGB1D2
Polyclonal
Unconjugated
SCGB1D2
LIPB; LIPHB; Lipophilin-B; LPHB; LPNB; prostatein-like lipophilin B; SCGB1D2; Secretoglobin family 1D member 2; secretoglobin, family 1D member 2; secretoglobin, family 1D, member 2
Rabbit
Antigen affinity chromatography
RUO
10647
-20°C
Liquid
Immunohistochemistry (Paraffin)
2.7 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
O95969
SCGB1D2
Synthetic peptide corresponding to residues near the C terminal of human secretoglobin, family 1D, member 2.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.