Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCGB1D2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557224
Description
Immunogen sequence: FCPALVSELL DFFFISEPLF KLSLAKFDAP PEAVAAKLGV KRCTDQMSLQ KRSLIAEVLV KILKKCS Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 45%.
May bind androgens and other steroids, may also bind estramustine, a chemotherapeutic agent used for prostate cancer. May be under transcriptional regulation of steroid hormones.
Specifications
| SCGB1D2 | |
| Polyclonal | |
| Unconjugated | |
| SCGB1D2 | |
| LIPB; LIPHB; Lipophilin-B; LPHB; LPNB; prostatein-like lipophilin B; SCGB1D2; Secretoglobin family 1D member 2; secretoglobin, family 1D member 2; secretoglobin, family 1D, member 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10647 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95969 | |
| SCGB1D2 | |
| Recombinant protein corresponding to Human SCGB1D2. Recombinant protein control fragment (Product #RP-88759). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction