Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCGB3A1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987239
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7239 Supplier Bioss Supplier No. BS6397R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Potential growth inhibitory cytokine. Classified as a secretoglobin; may have a role in airway function.

Specifications

Antigen SCGB3A1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SCGB3A1
Gene Accession No. Q96QR1
Gene Alias cytokine high in normal-1; Cytokine HIN-1; High in normal 1; HIN1; HIN-1; LU105; Pneumo secretory protein 2; PNSP2; PnSP-2; SCGB3A1; Secretoglobin family 3A member 1; secretoglobin, family 3A, member 1; UGRP2; UGRP2MGC87867; UNQ629/PRO1245; Uteroglobin-related protein 2
Gene Symbols SCGB3A1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SCGB3A1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92304
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.