Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCGF Mouse anti-Human, Clone: 4H11E4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | SCGF |
---|---|
Clone | 4H11E4 |
Concentration | 2 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6390
![]() ![]() |
Proteintech Group Inc
602951IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a member of the C-type lectin superfamily. The encoded protein is a secreted sulfated glycoprotein and functions as a growth factor for primitive hematopoietic progenitor cells. An alternative splice variant has been described but its biological nature has not been determined.Specifications
SCGF | |
2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CLEC11A, CLECSF3, LSLCL, P47, SCGF, Stem cell growth factor | |
CLEC11A | |
IgG2b | |
Protein A | |
Antibody |
4H11E4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9Y240 | |
6320 | |
SCGF Fusion Protein Ag21365 | |
Primary | |
-20°C | |
CLEC11A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title