Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCGN Monoclonal Antibody (CL0273)
GREENER_CHOICE

Catalog No. PIMA531338
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531338 Supplier Invitrogen™ Supplier No. MA531338
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: RDLFLHHKKA ISEAKLEEYT GTMMKIFDRN KDGRLDLNDL ARILALQENF LLQFKMDACS TEERKRDFEK IFAYYDVSKT GALEGPEVDG FVKDMMELVQ PSISGVDLDK FREILLRHCD VNKDGKIQKS ELALCLGLK Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.

The encoded protein is a secreted calcium-binding protein which is found in the cytoplasm. It is related to calbindin D-28K and calretinin. This protein is thought to be involved in KCL-stimulated calcium flux and cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCGN
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL0273
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCGN
Gene Accession No. O76038
Gene Alias CALBL; DJ501N12.8; SCGN; SECRET; Secretagogin; secretagogin, EF-hand calcium binding protein; SEGN; setagin
Gene Symbols SCGN
Host Species Mouse
Immunogen Recombinant protein fragment of Human SCGN (Product #RP-102381)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10590
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.