Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCGN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143619
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143619 100 μL
PIPA5143521 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5143619 Supplier Invitrogen™ Supplier No. PA5143619
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCGN Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)

The encoded protein is a secreted calcium-binding protein which is found in the cytoplasm. It is related to calbindin D-28K and calretinin. This protein is thought to be involved in KCL-stimulated calcium flux and cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCGN
Applications Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5mM sodium azide
Gene SCGN
Gene Accession No. O76038, Q6R556, Q91WD9
Gene Alias CALBL; DJ501N12.8; SCGN; SECRET; Secretagogin; secretagogin, EF-hand calcium binding protein; SEGN; setagin
Gene Symbols SCGN
Host Species Rabbit
Immunogen Full-length recombinant human secretagogin protein expressed in and purified from E. coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10590, 214189, 306942
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.