Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCGN Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530392
Description
Recommended positive controls: A549, HeLa. Predicted reactivity: Pig (84%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The encoded protein is a secreted calcium-binding protein which is found in the cytoplasm. It is related to calbindin D-28K and calretinin. This protein is thought to be involved in KCL-stimulated calcium flux and cell proliferation.
Specifications
| SCGN | |
| Polyclonal | |
| Unconjugated | |
| SCGN | |
| CALBL; DJ501N12.8; SCGN; SECRET; Secretagogin; secretagogin, EF-hand calcium binding protein; SEGN; setagin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10590 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.79 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O76038 | |
| SCGN | |
| Recombinant protein encompassing a sequence within the center region of human SCGN. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction