Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SCH 202676 hydrobromide
GREENER_CHOICE

Catalog No. 140010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-001-0 10 mg
14-005-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-001-0 Supplier Tocris Bioscience™ Supplier No. 1400/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibitor of ligand binding to G-protein-coupled receptors

Sulphydryl-reactive compound that inhibits agonist and antagonist binding to G-protein-coupled receptors. Inhibits a variety of GPCRs including adenosine, opioid, muscarinic, adrenergic and dopaminergic receptors (IC50 values are 0.1-1.8 μM).

Chemical Identifiers

CAS 265980-25-4
Molecular Formula C15H14BrN3S

Specifications

Quantity 10 mg
Formula Weight 348.26
Purity >99%
Molecular Weight (g/mol) 348.262
Chemical Name or Material SCH 202676 hydrobromide
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.