Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SCH 336
GREENER_CHOICESDP

Catalog No. 581550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-155-0 50 mg
58-151-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 58-155-0 Supplier Tocris Bioscience™ Supplier No. 5815/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly potent and selective CB2 inverse agonist

SCH 336 is a highly potent and selective CB2 receptor inverse agonist (Ki = 1.8 nM, EC50 = 2 nM). Displays 100-fold selectivity for CB2 receptors over CB1. Increases forskolin stimulated cAMP accumulation in CHO cells expressing human CB2 receptors. Inhibits leukocyte migration in a murine model of delayed-type hypersensitivity and inhibits antigen-induced lung eosinophilia in a mouse allergy model.

Specifications

Chemical Name or Material N-[(1S)-1-[4-[[4-Methoxy-2-[(4-methoxyphenyl)sulfonyl]phenyl]sulfonyl]phenyl]ethyl]methanesulfonamide
CAS 447459-51-0
Assay Percent Range >98%
Quantity 50 mg
Molecular Formula C23H25NO8S3
Formula Weight Observed MW: 539.64
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.