Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SCH 442416
GREENER_CHOICE

Catalog No. 24631
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-631 1 mg
24-631-0 10 mg
24-635-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 24-631 Supplier Tocris Bioscience™ Supplier No. 2463/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Very selective, high affinity A2A antagonist

Selective adenosine A2A receptor antagonist; binds to human and rat A2A receptors with high affinity (Ki values are 0.048 and 0.5 nM respectively). Displays > 23000-fold selectivity for hA2A over hA1 in vitro with minimal affinity for hA2B and hA3 receptors (IC50 > 10 μM). Blocks the cytoprotective effect of A2A agonist CGS-21680 (Cat.No. 1063).

Chemical Identifiers

CAS 316173-57-6
Molecular Formula C20H19N7O2

Specifications

Quantity 1 mg
Formula Weight 389.42
Purity >99%
Molecular Weight (g/mol) 389.419
Chemical Name or Material SCH 442416
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.