Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SCH 58261
GREENER_CHOICESDP

Catalog No. 227010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-701-0 10 mg
22-705-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-701-0 Supplier Tocris Bioscience™ Supplier No. 2270/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, highly selective A2A antagonist

SCH 58261 is a potent and selective A2A adenosine receptor competitive antagonist (Ki = 1.3 nM). Displays 323-, 53- and 100-fold selectivity over A1, A2B and A3 receptors, respectively. Reduces melanoma tumor growth and TGF-β levels in WT and P2X7R-null mice.

Chemical Identifiers

CAS 160098-96-4
Molecular Formula C18H15N7O

Specifications

Quantity 10 mg
Formula Weight 345.36
Purity >99%
Molecular Weight (g/mol) 345.366
Chemical Name or Material SCH 58261
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.