Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SCH 79797 dihydrochloride
GSA_VAGREENER_CHOICE

Catalog No. 159210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-921-0 10 mg
15-925-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-921-0 Supplier Tocris Bioscience™ Supplier No. 1592/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective non-peptide PAR1 antagonist

Potent, selective non-peptide PAR1 receptor antagonist (IC50 = 70 nM). Inhibits haTRAP-induced- but not γ-thrombin-, ADP- or collagen-induced human platelet aggregation. Also selectively blocks PAR1 agonist- or thrombin-induced increases in cytosolic Ca2+ in vascular smooth muscle cells.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 1216720-69-2
Molecular Formula C23H27Cl2N5

Specifications

Quantity 10 mg
Formula Weight 444.41
Purity >99%
Molecular Weight (g/mol) 444.404
Chemical Name or Material SCH 79797 dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.