Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Schiff's reagent

Catalog No. p-7078317 Shop All MilliporeSigma Products
Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass Bottle
Plastic Bottle

Catalog No. M1090330500

Supplier: MilliporeSigma™ 1.09033.0500

Only null left
Add to Cart
Add to Cart

Used for Microscopy

  • Used for detection of mucopolysaccharide, glycogen, muco, glycoproteins, glyco, phospholipids, basal membranes collagen
  • Schiff's reagent aldehydes give bright red color reaction nuclei appear blue
TRUSTED_SUSTAINABILITY

Specifications

Packaging Plastic Bottle
Quantity 500mL
Storage Requirements 15° to 25°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.