Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCHIP1 Antibody (OTI1C8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274008H
Description
SCHIP1 Monoclonal specifically detects SCHIP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SCHIP1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
SCHIP1 | |
Full length recombinant protein of human SCHIP1 (NP_055390) produced in HEK293T cells. | |
0.1 mL | |
29970.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1C8 | |
Western Blot, Immunohistochemistry | |
FLJ39160, SCHIP-1schwannomin-interacting protein 1, schwannomin interacting protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction