Learn More
Invitrogen™ SCHIP1 Monoclonal Antibody (OTI1C8)

Description
SCHIP1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | SCHIP1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1C8 |
| Concentration | 0.62 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Schip1 |
| Gene Accession No. | P0DPB3 |
| Gene Alias | hypothetical protein LOC550605; LOC478678; MGC128591 protein; neurofibromatosis 2 interacting protein; Nf2ip; Schip1; SCHIP-1; schwannomin interacting protein 1; schwannomin-interacting protein 1; zgc:110585 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.