Learn More
SCHIP1 Mouse anti-Human, Clone: OTI1A6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504440

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This locus represents naturally occurring read-through transcription from the neighboring IQ motif containing J (IQCJ) and schwannomin interacting protein 1 (SCHIP1) genes. Alternative splicing results in multiple transcript variants that are composed of in-frame exons from each individual gene. The resulting fusion products are thought to be components of the multimolecular complexes of axon initial segments and nodes of Ranvier, and they may play a role in calcium-mediated responses.Specifications
SCHIP1 | |
Monoclonal | |
Unconjugated | |
SCHIP1 | |
SCHIP-1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
29970 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1A6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9P0W5 | |
SCHIP1 | |
Full length human recombit protein of human SCHIP1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.