Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IQCJ-SCHIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551937
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551937 Supplier Invitrogen™ Supplier No. PA551937
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SNSTKVTPCS ECKSSSSPGG SLDLVSALED YEEPFPVYQK KVIDEWAPEE DGEEEEEEDE RDQRGYRDDR SPAREPGDVS ARTRSGGGGG RSATTAMPPP VPNGNLHQHD PQDLRHNGNV VV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 85%.

This locus represents naturally occurring read-through transcription from the neighboring IQ motif containing J (IQCJ) and schwannomin interacting protein 1 (SCHIP1) genes. Alternative splicing results in multiple transcript variants that are composed of in-frame exons from each individual gene. The resulting fusion products are thought to be components of the multimolecular complexes of axon initial segments and nodes of Ranvier, and they may play a role in calcium-mediated responses. [provided by RefSeq, Oct 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen IQCJ-SCHIP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IQCJ-SCHIP1
Gene Accession No. P0DPB3
Gene Alias IQCJ; SCHIP1
Gene Symbols IQCJ-SCHIP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCHIP1. Recombinant protein control fragment (Product #RP-101063).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100505385
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.