Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Schisandrin A, Thermo Scientific Chemicals

Catalog No. AC465030010
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61281-38-7
Molecular Weight (g/mol) 416.51
InChI Key JEJFTTRHGBKKEI-UHFFFAOYNA-N
IUPAC Name 3,4,5,14,15,16-hexamethoxy-9,10-dimethyltricyclo[10.4.0.0²,⁷]hexadeca-1(12),2,4,6,13,15-hexaene
SMILES COC1=CC2=C(C(OC)=C1OC)C1=C(OC)C(OC)=C(OC)C=C1CC(C)C(C)C2

Specifications

Molecular Formula C24H32O6
Quantity 1 g
Formula Weight 416.51
Chemical Name or Material Schisandrin A

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.