Learn More
Stoelting™ Schwartz T-Clip
SDP
Stoelting™ Schwartz T-Clips provide a very easy and secure method of stopping blood flow while performing procedures on vessels temporarily.
$78.40
Specifications
For Use With (Application) Neuroscience
Size 1 in.
View More Specs
Products 3
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-001-259
SDP
View Documents
Stoelting™
5210674
Straight, Smooth
Each for $78.40
Only null left
Add to Cart
10-001-260
SDP
View Documents
Stoelting™
5210675
Slight angle
Each for $78.40
Only null left
Add to Cart
10-001-261
SDP
View Documents
Stoelting™
5210676
Strong angle
Each for $78.40
Only null left
Add to Cart
Description
Description
  • Using the applying forceps, 52106-73, surgical clips can easily be position on the vessels. The locking forceps allows for easy, precise, and secure grasping of the micro surgical clips. Simply grasp the clip with the forceps to open the jaw, slid it over the vessels and release.
  • To remove surgical clips, simply perform the procedure in reverse.
  • This procedure does not compromise the vessels integrity.
Specifications
Specifications
Neuroscience
1 in.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.