Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Science Cube Kit - Just Tubes
GREENER_CHOICESCI_ED

Catalog No. S89524 Shop All Supertek Scientific Products
Click to view available options
Grade:
1
This item is not returnable. View return policy
This item is not returnable. View return policy

Just Tubes is an assortment of 3 different types of test tubes: glass, plastic, and safety. Test tubes are one of the most widely used tools for performing science experiments. Test tubes have applications in chemistry and biology.

  • Perfect for Group Experiments
  • Perfect for Homeschooling
  • Save space when storing in reusable plastic snap box
  • Includes:
    • 1 Reusable plastic snap box
    • 6 glass test tubes
    • 6 plastic test tubes
    • 6 safety test tubes
    • 1 Exploration Guide
  • Ages 7+

Specifications

Grade 1
Age 7
Includes Glass, Plastic, and Safety Test Tubes
Class Size 1 Lab Group
Product Type Supertek™ Science Cube Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.