Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Science Cube Kit - Test Tubes, Rack + More

Shop All Supertek Scientific Products


Click to view available options
Material:
Borosilicate Glass
Polystyrene
Description
- Perfect for Group Experiments
- Perfect for Homeschooling
- Save space when storing in reusable plastic snap box
- Includes:
- 1 Reusable plastic snap box
- 6 Test Tubes
- 2 Pipettes
- 1 Test Tube Tong
- 2 Test Tube Brushes
- 1 Test Tube Holder
- 1 Exploration Guide
- Ages 7+
Specifications
Specifications
Grade | 1 |
Age | 7 |
Material | Polystyrene |
Class Size | 1 Lab Group |
Product Type | Supertek™ Science Cube Kits |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction