Learn More
Science First™ Conductivity Meter
Description
This 10 level meter visually compares conductivity between different solutions. The green LED, numbered from 1-10 will light up as the probes are dipped into solutions of salts or acids.
- 10 Level
- Green LED
Specifications
Specifications
| For Use With (Application) | Conductivity |
| Display Type | LED |
| Sensor Types | Conductivity |