Learn More
Science First™ Variable Induction Kit
SDPSCI_ED
Catalog No. S98906
Click to view available options
:
Changing currents demo
Catalog No. S98906
Supplier: Science First™ 6154130
Current induced into a circuit is constantly changing and this effect is most notable for its use in RF transmitters.
  • Using the hand crank, slide a steel core in and out of a copper coil to induce varying amounts of current
  • Variable Inductance apparatus are unusual and hard to find pieces for the lab
Specifications
Product Type Variable Induction Kit
For Use With (Equipment) RF transmitters
For Use With (Application) To study the phenomenon of changing currents induced into a circuit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.